2-(4,5-Dimethylthiazol-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

498620-5G 498620-25G 498620-1G 498620-500MG
77630-082EA 1484.23 USD
77630-082 77659-840 77643-458 77647-420
2-(4,5-Dimethylthiazol-2-yl)propan-2-ol ≥95%
2-(4,5-Dimethylthiazol-2-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 171.26 g/mol
MW: C₈H₁₃NOS
Storage Temperature: Ambient
CAS Number: 1517530-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR