1-(4,5-Dimethylthiazol-2-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

498605-5G 498605-1G 498605-500MG
77630-080EA 1484.23 USD
77630-080 77647-418 77651-398
1-(4,5-Dimethylthiazol-2-yl)propan-1-one ≥95%
1-(4,5-Dimethylthiazol-2-yl)propan-1-one

Precaución: For Research Use Only

Formula: 169.244 g/mol
MW: C₈H₁₁NOS
Storage Temperature: Ambient
CAS Number: 2172006-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR