4,5-Dimethyl-2-(methylthio)thiazole ≥95%

Supplier: AOBChem USA

Warning

498603-5G 498603-1G 498603-25G 498603-500MG
77633-454EA 1381 USD
77633-454 77639-424 77659-838 77651-396
4,5-Dimethyl-2-(methylthio)thiazole ≥95%
4,5-Dimethyl-2-(methylthio)thiazole

Precaución: For Research Use Only

Formula: 159.272 g/mol
MW: C₆H₉NS₂
Storage Temperature: Ambient
CAS Number: 17626-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR