1-Bromo-5-chloro-3-methoxy-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

498447-5G 498447-25G 498447-1G 498447-500MG
77635-114EA 298 USD
77635-114 77656-300 77643-454 77651-394
1-Bromo-5-chloro-3-methoxy-2-(methoxymethoxy)benzene ≥95%
1-Bromo-5-chloro-3-methoxy-2-(methoxymethoxy)benzene

Precaución: For Research Use Only

Formula: 281.5309 g/mol
MW: C₉H₁₀BrClO₃
Storage Temperature: Ambient
CAS Number: 3037682-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR