Tert-butyl 4-chloro-2-ethoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

498022-5G 498022-25G 498022-1G 498022-500MG
77630-072EA 1484.23 USD
77630-072 77636-028 77639-418 77647-410
Tert-butyl 4-chloro-2-ethoxybenzoate ≥95%
Tert-butyl 4-chloro-2-ethoxybenzoate

Precaución: For Research Use Only

Formula: 256.725 g/mol
MW: C₁₃H₁₇ClO₃
Storage Temperature: Ambient
CAS Number: 3031789-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR