1-(2,4-Difluoro-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

497836-5G 497836-25G 497836-1G 497836-500MG
77630-070EA 1484.23 USD
77630-070 77636-026 77643-450 77651-392
1-(2,4-Difluoro-3-methylphenyl)cyclobutanol ≥95%
1-(2,4-Difluoro-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
CAS Number: 3031791-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR