5-Methylthiophene-2-carbaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

497573-5G 497573-25G 497573-1G 497573-500MG
77633-452EA 844 USD
77633-452 77656-298 77643-448 77647-408
5-Methylthiophene-2-carbaldehyde oxime ≥97%
5-Methylthiophene-2-carbaldehyde oxime

Precaución: For Research Use Only

Formula: C₆H₇NOS
MW: 141.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01197328
CAS Number: 1887033-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR