Tetrahydrothiophen-3-ol ≥97%

Supplier: AOBChem USA

Warning

493376-5G 493376-25G 493376-1G 493376-500MG
77635-108EA 256 USD
77635-108 77659-820 77643-438 77647-392
Tetrahydrothiophen-3-ol ≥97%
Tetrahydrothiophen-3-ol

Precaución: For Research Use Only

Formula: C₄H₈OS
Storage Temperature: Ambient
MDL Number: MFCD00121060
CAS Number: 3334-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR