1-(5-Bromobenzo[b]thiophen-2-yl)butan-1-one ≥95%

Supplier: AOBChem USA

Warning

493372-5G 493372-25G 493372-1G 493372-500MG
77630-064EA 1484.23 USD
77630-064 77659-818 77643-436 77651-384
1-(5-Bromobenzo[b]thiophen-2-yl)butan-1-one ≥95%
1-(5-Bromobenzo[b]thiophen-2-yl)butan-1-one

Precaución: For Research Use Only

Formula: C₁₂H₁₁BrOS
Storage Temperature: Ambient
CAS Number: 3031805-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR