Ethyl 2-(3-isopropoxy-4-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

493297-5G 493297-1G 493297-500MG
77630-062EA 1484.23 USD
77630-062 77643-432 77647-388
Ethyl 2-(3-isopropoxy-4-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-isopropoxy-4-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 250.29 g/mol
MW: C₁₄H₁₈O₄
Storage Temperature: Ambient
CAS Number: 3031789-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR