2-(3-Methylthiophen-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

493294-5G 493294-1G 493294-500MG
77656-292EA 2250 USD
77656-292 77643-430 77647-386
2-(3-Methylthiophen-2-yl)propan-2-ol ≥95%
2-(3-Methylthiophen-2-yl)propan-2-ol

Precaución: For Research Use Only

Formula: 156.245 g/mol
MW: C₈H₁₂OS
Storage Temperature: Ambient
CAS Number: 1340552-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR