Trimethyl(4-methyl-5-(methylthio)thiophen-2-yl)silane ≥95%

Supplier: AOBChem USA

Warning

493233-5G 493233-25G 493233-1G 493233-500MG
77630-060EA 1484.23 USD
77630-060 77636-024 77639-412 77651-382
Trimethyl(4-methyl-5-(methylthio)thiophen-2-yl)silane ≥95%
Trimethyl(4-methyl-5-(methylthio)thiophen-2-yl)silane

Precaución: For Research Use Only

Formula: C₉H₁₆S₂Si
Storage Temperature: Ambient
CAS Number: 3037695-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR