1-(4-Methyl-5-(methylthio)thiophen-2-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

493232-5G 493232-1G 493232-25G 493232-500MG
77630-058EA 1484.23 USD
77630-058 77639-410 77656-290 77647-384
1-(4-Methyl-5-(methylthio)thiophen-2-yl)propan-1-one ≥95%
1-(4-Methyl-5-(methylthio)thiophen-2-yl)propan-1-one

Precaución: For Research Use Only

Formula: 200.321 g/mol
MW: C₉H₁₂OS₂
Storage Temperature: Ambient
CAS Number: 3037705-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR