5-Bromo-3-methyl-2-(methylthio)thiophene ≥95%

Supplier: AOBChem USA

Warning

493231-5G 493231-25G 493231-1G 493231-500MG
77630-056EA 1484.23 USD
77630-056 77636-022 77643-428 77651-380
5-Bromo-3-methyl-2-(methylthio)thiophene ≥95%
5-Bromo-3-methyl-2-(methylthio)thiophene

Precaución: For Research Use Only

Formula: 223.154 g/mol
MW: C₆H₇BrS₂
Storage Temperature: Ambient
CAS Number: 3037703-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR