1-(4-Methyl-5-(methylthio)thiophen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

493229-5G 493229-25G 493229-1G 493229-500MG
77630-052EA 1484.23 USD
77630-052 77659-814 77643-426 77647-382
1-(4-Methyl-5-(methylthio)thiophen-2-yl)ethanone ≥95%
1-(4-Methyl-5-(methylthio)thiophen-2-yl)ethanone

Precaución: For Research Use Only

Formula: 186.294 g/mol
MW: C₈H₁₀OS₂
Storage Temperature: Ambient
CAS Number: 3037703-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR