1-(2,3-Dichlorophenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

492610-5G 492610-25G 492610-1G 492610-500MG
77633-434EA 1034 USD
77633-434 77656-280 77643-420 77651-372
1-(2,3-Dichlorophenyl)cyclopentanol ≥95%
1-(2,3-Dichlorophenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂Cl₂O
Storage Temperature: Ambient
CAS Number: 1343082-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR