1-(5-Ethoxy-2-methylphenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

492537-5G 492537-1G 492537-500MG
77630-032EA 1484.23 USD
77630-032 77639-390 77651-370
1-(5-Ethoxy-2-methylphenyl)-2-methoxyethanone ≥95%
1-(5-Ethoxy-2-methylphenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 208.254 g/mol
MW: C₁₂H₁₆O₃
Storage Temperature: Ambient
CAS Number: 3031807-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR