1-(5-Ethoxy-2-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

492536-5G 492536-1G 492536-25G 492536-500MG
77630-030EA 1484.23 USD
77630-030 77639-388 77659-804 77651-368
1-(5-Ethoxy-2-methylphenyl)-2,2-difluoroethanone ≥95%
1-(5-Ethoxy-2-methylphenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: 214.209 g/mol
MW: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
CAS Number: 3031785-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR