2-Chloro-1-(5-ethoxy-2-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

492535-5G 492535-1G 492535-25G 492535-500MG
77630-028EA 1484.23 USD
77630-028 77639-386 77656-274 77651-366
2-Chloro-1-(5-ethoxy-2-methylphenyl)ethanone ≥95%
2-Chloro-1-(5-ethoxy-2-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 212.673 g/mol
MW: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
CAS Number: 3031789-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR