(5-Ethoxy-2-methylphenyl)trimethylsilane ≥95%
Supplier: AOBChem USA
|
Warning
|
492525-5G
492525-25G
492525-1G
492525-500MG
77630-016EA
1484.23
USD
77630-016
77656-272
77643-408
77647-360
(5-Ethoxy-2-methylphenyl)trimethylsilane ≥95%
(5-Ethoxy-2-methylphenyl)trimethylsilane
Precaución: For Research Use Only
Formula:
208.372 g/mol MW: C₁₂H₂₀OSi Storage Temperature: Ambient |
CAS Number:
3031805-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...