2-(3,4-Dibromothiophen-2-yl)propan-2-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
492509-5G
492509-1G
492509-25G
492509-500MG
77630-012EA
1484.23
USD
77630-012
77639-380
77659-798
77651-360
2-(3,4-Dibromothiophen-2-yl)propan-2-ol ≥97%
2-(3,4-Dibromothiophen-2-yl)propan-2-ol
Precaución: For Research Use Only
Formula:
300.011 g/mol MW: C₇H₈Br₂OS Storage Temperature: Ambient |
CAS Number:
3037676-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...