3-Bromo-6-methoxypicolinic acid ≥97%

Supplier: AOBChem USA

49233-5G 49233-25G
77635-106EA 80 USD
77635-106 77656-270
3-Bromo-6-methoxypicolinic acid ≥97%
3-Bromo-6-methoxypicolinic acid

Precaución: For Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD13185812
CAS Number: 1196147-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR