3-Chloro-2-fluoro-5-nitrobenzotrifluoride ≥97%

Supplier: AOBChem USA

Synonyms: 1-Chloro-2-fluoro-5-nitro-3-(trifluoromethyl)benzene

492196-5G 492196-25G
77635-104EA 77 USD
77635-104 77656-268
3-Chloro-2-fluoro-5-nitrobenzotrifluoride ≥97%
3-Chloro-2-fluoro-5-nitrobenzotrifluoride

Precaución: For Research Use Only

Formula: C₇H₂ClF₄NO₂
MW: 243.5429 g/mol
MDL Number: MFCD09033153
CAS Number: 914225-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR