(5-Ethoxy-2-methylphenyl)(methyl)sulphane ≥95%

Supplier: AOBChem USA

Warning

491890-5G 491890-25G 491890-1G 491890-500MG
77630-008EA 1484.23 USD
77630-008 77659-796 77643-402 77651-358
(5-Ethoxy-2-methylphenyl)(methyl)sulphane ≥95%
(5-Ethoxy-2-methylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
CAS Number: 3031798-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR