1-(3-Ethoxy-2-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

491821-5G 491821-25G 491821-1G 491821-500MG
77630-006EA 1484.23 USD
77630-006 77656-266 77643-400 77647-356
1-(3-Ethoxy-2-methylphenyl)cyclobutanol ≥95%
1-(3-Ethoxy-2-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3037705-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR