2-(3-Ethoxy-2-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

491820-5G 491820-1G 491820-25G 491820-500MG
77630-004EA 1484.23 USD
77630-004 77639-378 77656-264 77651-356
2-(3-Ethoxy-2-methylphenyl)propan-2-ol ≥95%
2-(3-Ethoxy-2-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 194.27 g/mol
MW: C₁₂H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3037705-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR