(7-Chloronaphthalen-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

491814-1G 491814-5G 491814-500MG
77639-374EA 628 USD
77639-374 77656-262 77651-354
(7-Chloronaphthalen-2-yl)boronic acid ≥97%
(7-Chloronaphthalen-2-yl)boronic acid

Precaución: For Research Use Only

Formula: 206.4333 g/mol
MW: C₁₀H₈BClO₂
Storage Temperature: Ambient
CAS Number: 2118958-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR