1-Ethoxy-2-methoxy-4-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

491787-5G 491787-25G 491787-1G 491787-500MG
77633-432EA 1425 USD
77633-432 77656-260 77643-398 77647-354
1-Ethoxy-2-methoxy-4-nitrobenzene ≥97%
1-Ethoxy-2-methoxy-4-nitrobenzene

Precaución: For Research Use Only

Formula: 197.1879 g/mol
MW: C₉H₁₁NO₄
Storage Temperature: Ambient
CAS Number: 7244-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR