3-(1,3-Dioxolan-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

491777-5G 491777-1G 491777-500MG
77630-002EA 1841.64 USD
77630-002 77639-372 77647-352
3-(1,3-Dioxolan-2-yl)benzaldehyde ≥97%
3-(1,3-Dioxolan-2-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
CAS Number: 68348-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR