Methyl 2,6-difluoro-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

49058-5G 49058-1G 49058-25G 49058-500MG
77633-430EA 1070 USD
77633-430 77639-370 77659-788 77651-352
Methyl 2,6-difluoro-3-methylbenzoate ≥95%
Methyl 2,6-difluoro-3-methylbenzoate

Precaución: For Research Use Only

Formula: 186.155 g/mol
MW: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06203779
CAS Number: 773873-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR