4-Bromo-5-chloro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

490224-5G 490224-1G 490224-25G 490224-500MG
77635-094EA 570 USD
77635-094 77639-366 77659-784 77651-348
4-Bromo-5-chloro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95%
4-Bromo-5-chloro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: For Research Use Only

Formula: 315.59 g/mol
MW: C₁₂H₁₂BrClN₂O
Storage Temperature: Ambient
CAS Number: 2158302-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR