5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

490124-5G 490124-25G 490124-1G 490124-500MG
77633-428EA 1282 USD
77633-428 77659-782 77643-394 77651-346
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥97%
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: For Research Use Only

Formula: C₁₂H₁₃IN₂O
Storage Temperature: Ambient
CAS Number: 1337882-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR