2-Fluoro-α,α,3-trimethylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

489590-5G 489590-1G 489590-25G 489590-500MG
77633-426EA 1171 USD
77633-426 77639-364 77656-252 77647-348
2-Fluoro-α,α,3-trimethylbenzenemethanol ≥95%
2-Fluoro-α,α,3-trimethylbenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₃FO
Storage Temperature: Ambient
CAS Number: 1522537-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR