(4-Methyl-2-(methylthio)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

489221-5G 489221-25G 489221-1G 489221-500MG
77629-992EA 1484.23 USD
77629-992 77656-244 77643-384 77647-342
(4-Methyl-2-(methylthio)phenyl)(phenyl)methanone ≥95%
(4-Methyl-2-(methylthio)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₅H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037696-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR