1-(4-Methyl-2-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

489220-5G 489220-500MG 489220-25G 489220-1G
77629-990EA 1484.23 USD
77629-990 77652-944 77659-780 77643-382
1-(4-Methyl-2-(methylthio)phenyl)propan-1-one ≥95%
1-(4-Methyl-2-(methylthio)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 194.293 g/mol
MW: C₁₁H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037703-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR