Cyclopropyl(4-methyl-2-(methylthio)phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

489219-5G 489219-25G 489219-1G 489219-500MG
77629-988EA 1484.23 USD
77629-988 77636-004 77639-358 77651-344
Cyclopropyl(4-methyl-2-(methylthio)phenyl)methanone ≥95%
Cyclopropyl(4-methyl-2-(methylthio)phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037703-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR