2,2,2-Trifluoro-1-(4-methyl-2-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

489218-5G 489218-25G 489218-1G 489218-500MG
77629-986EA 1484.23 USD
77629-986 77659-778 77643-380 77651-342
2,2,2-Trifluoro-1-(4-methyl-2-(methylthio)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-methyl-2-(methylthio)phenyl)ethanone

Precaución: For Research Use Only

Formula: 234.238 g/mol
MW: C₁₀H₉F₃OS
Storage Temperature: Ambient
CAS Number: 3037692-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR