Methyl 4-amino-5-bromopicolinate ≥97%

Supplier: AOBChem USA

Warning

489123-5G 489123-1G 489123-25G 489123-500MG
77635-092EA 528 USD
77635-092 77639-356 77656-242 77647-340
Methyl 4-amino-5-bromopicolinate ≥97%
Methyl 4-amino-5-bromopicolinate

Precaución: For Research Use Only

Formula: C₇H₇BrN₂O₂
Storage Temperature: Ambient
CAS Number: 1782230-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR