5-Bromo-2-(p-tolyl)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

489122-5G 489122-1G 489122-25G 489122-500MG
77633-420EA 728 USD
77633-420 77639-354 77656-240 77651-338
5-Bromo-2-(p-tolyl)pyrimidine ≥97%
5-Bromo-2-(p-tolyl)pyrimidine

Precaución: For Research Use Only

Formula: 249.107 g/mol
MW: C₁₁H₉BrN₂
Storage Temperature: Ambient
CAS Number: 1192164-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR