Tert-butyl 3-ethoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

489105-5G 489105-25G 489105-1G 489105-500MG
77629-982EA 1484.23 USD
77629-982 77636-002 77639-350 77647-338
Tert-butyl 3-ethoxy-5-methylbenzoate ≥97%
Tert-butyl 3-ethoxy-5-methylbenzoate

Precaución: For Research Use Only

Formula: 236.307 g/mol
MW: C₁₄H₂₀O₃
Storage Temperature: Ambient
CAS Number: 3031791-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR