1,2-Bis(4-methoxy-3,5-dimethylphenyl)ethane-1,2-dione ≥95%

Supplier: AOBChem USA

Warning

489081-5G 489081-25G 489081-1G 489081-500MG
77629-980EA 1484.23 USD
77629-980 77659-774 77643-376 77651-334
1,2-Bis(4-methoxy-3,5-dimethylphenyl)ethane-1,2-dione ≥95%
1,2-Bis(4-methoxy-3,5-dimethylphenyl)ethane-1,2-dione

Precaución: For Research Use Only

Formula: 326.3862 g/mol
MW: C₂₀H₂₂O₄
Storage Temperature: Ambient
CAS Number: 3031812-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR