2-Isopropyl-6-methylphenol ≥95%

Supplier: AOBChem USA

Warning

489079-1G 489079-5G 489079-500MG
77635-090EA 1083 USD
77635-090 77656-236 77651-332
2-Isopropyl-6-methylphenol ≥95%
2-Isopropyl-6-methylphenol

Precaución: For Research Use Only

Formula: 150.2175 g/mol
MW: C₁₀H₁₄O
Storage Temperature: Ambient
CAS Number: 3228-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR