Ethyl 2-((5-bromopyridin-2-yl)thio)acetate ≥97%

Supplier: AOBChem USA

Warning

489068-5G 489068-1G 489068-25G 489068-500MG
77633-418EA 1266 USD
77633-418 77639-348 77656-234 77651-330
Ethyl 2-((5-bromopyridin-2-yl)thio)acetate ≥97%
Ethyl 2-((5-bromopyridin-2-yl)thio)acetate

Precaución: For Research Use Only

Formula: 276.1502 g/mol
MW: C₉H₁₀BrNO₂S
Storage Temperature: Ambient
CAS Number: 1249973-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR