1-(2,4-Dichlorophenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

488613-5G 488613-25G 488613-1G 488613-500MG
77633-416EA 689 USD
77633-416 77656-232 77643-374 77651-328
1-(2,4-Dichlorophenyl)-2-methoxyethanone ≥95%
1-(2,4-Dichlorophenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
CAS Number: 1157166-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR