1-(3-Fluorophenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

488448-5G 488448-25G 488448-1G 488448-500MG
77633-414EA 1368 USD
77633-414 77656-230 77643-372 77651-326
1-(3-Fluorophenyl)-2-methoxyethanone ≥95%
1-(3-Fluorophenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 168.17 g/mol
MW: C₉H₉FO₂
Storage Temperature: Ambient
CAS Number: 925211-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR