1-(3-Chlorophenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

488413-5G 488413-1G 488413-25G 488413-500MG
77635-088EA 548 USD
77635-088 77639-344 77656-228 77651-324
1-(3-Chlorophenyl)-2-methoxyethanone ≥95%
1-(3-Chlorophenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
CAS Number: 247179-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR