1-Bromo-3-fluoro-2-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48802-5G 48802-1G 48802-25G 48802-500MG
77635-086EA 361 USD
77635-086 77639-342 77656-224 77647-334
1-Bromo-3-fluoro-2-iodo-4-methoxybenzene ≥97%
1-Bromo-3-fluoro-2-iodo-4-methoxybenzene

Precaución: For Research Use Only

Formula: C₇H₅BrFIO
Storage Temperature: Ambient
MDL Number: MFCD32902768
CAS Number: 2385126-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR