2-(2,4-Difluoro-3-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

48786-5G 48786-1G 48786-25G
77635-084EA 179 USD
77635-084 77639-340 77659-770
2-(2,4-Difluoro-3-methoxyphenyl)-1,3-dioxolane ≥95%
2-(2,4-Difluoro-3-methoxyphenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 216.181 g/mol
MW: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD32902764
CAS Number: 2586126-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR