(2-Fluoro-4-methoxy-3-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

487668-5G 487668-500MG 487668-25G 487668-1G
77633-410EA 1441 USD
77633-410 77652-936 77659-766 77643-356
(2-Fluoro-4-methoxy-3-methylphenyl)trimethylsilane ≥95%
(2-Fluoro-4-methoxy-3-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 212.336 g/mol
MW: C₁₁H₁₇FOSi
Storage Temperature: Ambient
CAS Number: 3037703-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR