3-(2-Methoxy-3,5-dimethylphenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

487591-5G 487591-1G 487591-500MG
77656-212EA 2338 USD
77656-212 77643-348 77651-310
3-(2-Methoxy-3,5-dimethylphenyl)oxetan-3-ol ≥95%
3-(2-Methoxy-3,5-dimethylphenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 208.254 g/mol
MW: C₁₂H₁₆O₃
Storage Temperature: Ambient
CAS Number: 1895392-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR